missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (5G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
209.00 € - 531.00 €
Specifications
| Antigen | Desmoglein-3 |
|---|---|
| Clone | 5G11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18740164
|
Novus Biologicals
NB100-1643SS |
0.025 mL |
209.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18716553
|
Novus Biologicals
NB100-1643 |
0.1 mL |
531.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot.Specifications
| Desmoglein-3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers, Extracellular Matrix, Signal Transduction | |
| P32926 | |
| 1830 | |
| Human Desmoglein 3 recombinant protein [UniProt# P32926] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5G11 | |
| Western Blot 1:500 - 1:1000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| DSG3 | |
| IgG1 | |
| Protein G purified | |
| This antibody reacts with Desmoglein-3. Does not cross-react with Desmoglein 1 or Desmoglein 2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title