missing translation for 'onlineSavingsMsg'
Learn More

Desoxycortone Acetate, Mikromol™

Product Code. 17970576
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970576 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970576 Supplier Mikromol Supplier No. MM1685.00
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Desoxycortone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deoxycorticosterone acetate
CAS 56-47-3
Analyte or Component Names Deoxycorticosterone acetate
Shipping Condition Room Temperature
Molecular Formula C23 H32 O4
Certifications/Compliance ISO 17034
Synonym Desoxycortone acetate, 3,20-Dioxopregn-4-en-21-yl acetate, 11-Deoxycorticosterone, acetate, Pregn-4-ene-3,20-dione, 21-hydroxy-, acetate, 11-Deoxycorticosterone 21-acetate, 11-Desoxycorticosterone acetate, 21-Acetoxy-3,20-diketopregn-4-ene, 21-Acetoxypregn-4-en-3,20-dione, 21-Acetoxypregn-4-ene-3,20-dione, 21-Hydroxypregn-4-ene-3,20-dione 21-acetate, 21-Hydroxypregn-4-ene-3,20-dione acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnene-3,20-dione 21-acetate, 4-Pregnene-3,20-dione-21-ol acetate, Arcort, Bio-Corten, Cortacet, Cortate, Cortenil, Cortesan, Cortexone acetate, Cortifar, Cortigen, Cortinaq, Cortiron, Cortivis, Cortixyl, DCA, DOC acetate, DOCA, Decorten, Decortin, Decosteron, Decosterone, Decostrate, Deoxycorticosterone 21-acetate, Deoxycorticosterone acetate, Descorterone, Descotone, Desoxycorticosterone acetate, Desoxycortone acetate, Dorcostrin, Doxo, Doxycamon, Krinocorts, NSC 9567, Ocriten, Ocritena, Percorten, Percotol, Primocort, Primocortan, Sincortex, Steraq, Syncort, Syncorta, Syncortyl, Unidocan
InChI Formula InChI=1S/C23H32O4/c1-14(24)27-13-21(26)20-7-6-18-17-5-4-15-12-16(25)8-10-22(15,2)19(17)9-11-23(18,20)3/h12,17-20H,4-11,13H2,1-3H3/t17-,18-,19-,20+,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [2-[(8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 372.5
Quantity 250 mg
Formula Weight 372.2301 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.