missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone EP Impurity D (Mometasone Furoate EP Impurity L), Mikromol™

Product Code. 30464103
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464103 4 x 25 mg -
30051100 100 mg 100mg
30052740 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464103 Supplier Mikromol Supplier No. MM0213.0400254AMP
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexamethasone EP Impurity D (Mometasone Furoate EP Impurity L), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione
CAS 24916-90-3
Analyte or Component Names 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione
Shipping Condition Room Temperature
Molecular Formula C22 H28 O5
Certifications/Compliance ISO 17025
Synonym 9β,11β-Epoxy-17,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Dexamethasone Imp. D (EP), 17,21-Dihydroxy-16alpha-methyl-9beta,11beta-epoxypregna-1,4-diene-3,20-dione, 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Dexamethasone 9,11-Epoxide, Dexamethasone Impurity D, Mometasone Furoate Impurity L
InChI Formula InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19+,20+,21+,22-/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione
Molecular Weight (g/mol) 372.45
Quantity 4 x 25 mg
Formula Weight 372.1937 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.