missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone EP Impurity E, Mikromol

Product Code. 30462995
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462995 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462995 Supplier Mikromol Supplier No. MM0213.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexamethasone EP Impurity E, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17,21-Dihydroxy-16alpha-methylpregna-1,4,9(11)-triene-3,20-dione
CAS 13209-41-1
Analyte or Component Names 17,21-Dihydroxy-16alpha-methylpregna-1,4,9(11)-triene-3,20-dione
Shipping Condition Room Temperature
Molecular Formula C22 H28 O4
Certifications/Compliance ISO 9001
Synonym 17,21-Dihydroxy-16α-methylpregna-1,4,9(11)-triene-3,20-dione, 17,21-Dihydroxy-16α-methyl-pregna-1,4,9(11)-triene-3,20-dione, Pregna-1,4,9(11)-triene-3,20-dione, 17,21-dihydroxy-16α-methyl- (6CI,7CI,8CI), 17α,21-Dihydroxy-16α-methylpregna-1,4,9(11)-triene-3,20-dione, VBP 15, Dexamethasone Impurity E
InChI Formula InChI=1S/C22H28O4/c1-13-10-18-16-5-4-14-11-15(24)6-8-20(14,2)17(16)7-9-21(18,3)22(13,26)19(25)12-23/h6-8,11,13,16,18,23,26H,4-5,9-10,12H2,1-3H3/t13-,16-,18+,20+,21+,22+/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name (8S,10S,13S,14S,16R,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 356.46
Quantity 50 mg
Formula Weight 356.1988 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.