missing translation for 'onlineSavingsMsg'
Learn More

Dexketoprofen, Mikromol™

Product Code. 18081750
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081750 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081750 Supplier Mikromol Supplier No. MM3535.08
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexketoprofen
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(+)-Ketoprofen
CAS 22161-81-5
Analyte or Component Names (S)-(+)-Ketoprofen
Shipping Condition Room Temperature
Molecular Formula C16 H14 O3
Certifications/Compliance ISO 17034
Synonym Hydratropic acid, m-benzoyl-, (+)- (8CI), (+)-(S)-m-Benzoylhydratropic acid, (+)-2-(3-Benzoylphenyl)propionic acid, (+)-3-Benzoylhydratropic acid, (+)-Ketoprofen, (2S)-2-(3-Benzoylphenyl)propionic acid, (S)-(+)-2-(3-Benzoylphenyl)propionic acid, (S)-2-(3-Benzoylphenyl)propanoic acid, (S)-2-(3-Benzoylphenyl)propionic acid, (S)-Ketoprofen, Arveles, Dexketoprofen, S-(+)-Ketoprofen, (alphaS)-3-benzoyl-alpha-methylbenzeneacetic acid, Benzeneacetic acid, 3-benzoyl-alpha-methyl-, (S)-, (+)-3-Benzoyl-alpha-methylbenzeneacetic acid, (alphaS)-3-Benzoyl-alpha-methylbenzeneacetic acid
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/t11-/m0/s1
SMILES C[C@H](C(=O)O)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name (2S)-2-(3-benzoylphenyl)propanoic acid
Molecular Weight (g/mol) 254.28
Quantity 250 mg
Formula Weight 254.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.