missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexmedetomidine Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Dexmedetomidine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Dexmedetomidine Hydrochloride |
| CAS | 145108-58-3 |
| Analyte or Component Names | Dexmedetomidine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 N2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dexmedetomidine Hydrochloride |
| InChI Formula | InChI=1S/C13H16N2.ClH/c1-9-5-4-6-12(10(9)2)11(3)13-7-14-8-15-13;/h4-8,11H,1-3H3,(H,14,15);1H/t11-;/m0./s1 |
| SMILES | Cl.C[C@H](c1c[nH]cn1)c2cccc(C)c2C |
| IUPAC Name | 4-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction