missing translation for 'onlineSavingsMsg'
Learn More

Dextromethorphan Hydrobromide Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal

Product Code. 30463076
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463076 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463076 Supplier LoGiCal Supplier No. LGCAMP0033.0001
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Dextromethorphan Hydrobromide Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dextromethorphan hydrobromide hydrate
CAS 6700-34-1
Analyte or Component Names Dextromethorphan hydrobromide hydrate
Shipping Condition Room Temperature
Molecular Formula C18 H25 N O . Br H . H2 O
Certifications/Compliance ISO 17034
Synonym Dextromethorphan Hydrobromide Monohydrate, Morphinan, 3-methoxy-17-methyl-, hydrobromide, hydrate (1:1:1), (9 alpha,13 alpha,14 alpha)-, 9 alpha,13 alpha,14 alpha-Morphinan, 3-methoxy-17-methyl-, hydrobromide, monohydrate (8 CI), Morphinan, 3-methoxy-17-methyl-, hydrobromide, monohydrate, (9 alpha,13 alpha,14 alpha)- (9 CI), (+)-3-Methoxy-N-methylmorphinan hydrobromide monohydrate, d-3-Methoxy-N-methylmorphinan hydrobromide monohydrate
InChI Formula InChI=1 S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17 H,3-5,8-11H2,1-2H3;1 H;1H2/t15-,17+,18+;;/m1../s1
SMILES O.Br.COc1ccc2C[C@H]3 [C@H]4 CCCC[C@@]4(CCN3C)c2c1
IUPAC Name (9α,13α,14α)-3-Methoxy-17-methylmorphinan hydrobromide monohydrate
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 370.324
Quantity 1 mL
Formula Weight 369.13 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.