missing translation for 'onlineSavingsMsg'
Learn More

DGK-alpha Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™

Product Code. 30491937 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30491937 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30491937 Supplier Novus Biologicals Supplier No. NBP335405MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DGK-alpha Polyclonal antibody specifically detects DGK-alpha in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGK-alpha
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human DGK-alpha (NP_001336.2).,, Sequence:, MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1606
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.