missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,f][1,4]thiazepin-11(10H)-one, Mikromol™

Product Code. 30049611
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049611 25 mg 25mg
17962606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049611 Supplier Mikromol Supplier No. MM1130.090025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dibenzo[b,f][1,4]thiazepin-11(10H)-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzo[b,f][1,4]thiazepin-11(10H)-one
CAS 7-7-3159
Analyte or Component Names Dibenzo[b,f][1,4]thiazepin-11(10H)-one
Shipping Condition Room Temperature
Molecular Formula C13 H9 N O S
Certifications/Compliance ISO 17025
Synonym Dibenzo[b,f][1,4]thiazepin-11(10H)-one, Quetiapine Fumarate Imp. G (EP), Quetiapine Related Compound G, Quetiapine Fumarate Impurity G, Quetiapine Impurity G
InChI Formula InChI=1S/C13H9NOS/c15-13-9-5-1-3-7-11(9)16-12-8-4-2-6-10(12)14-13/h1-8H,(H,14,15)
SMILES O=C1Nc2ccccc2Sc3ccccc13
IUPAC Name 5H-benzo[b][1,4]benzothiazepin-6-one
Molecular Weight (g/mol) 227.28
Quantity 25 mg
Formula Weight 227.0405 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.