missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Ether, Mikromol™

Product Code. 30051572
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051572 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051572 Supplier Mikromol Supplier No. MM1460.01
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dibenzyl Ether
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl ether
CAS 103-50-4
Analyte or Component Names Benzyl ether
Shipping Condition Room Temperature
Molecular Formula C14 H14 O
Certifications/Compliance ISO 17025
Synonym Tribenoside Imp. D (EP), 1,1'-[Oxybis(methylene)]bis-benzene, Benzyl ether, 1,1'-[Oxybis(methylene)]bis[benzene], BA, BA (plasticizer), Benzyl oxide, Dibenzyl ether, NSC 5931, Plastikator BA, Tribenoside Impurity D
InChI Formula InChI=1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2
SMILES C(OCc1ccccc1)c2ccccc2
IUPAC Name phenylmethoxymethylbenzene
Molecular Weight (g/mol) 198.26
Quantity 100 mg
Formula Weight 198.1045 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.