missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sebacate, Mikromol™

Product Code. 17920035
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
17920035 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 17920035

Brand: Mikromol MM0925.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dibutyl Sebacate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl Sebacate
CAS 109-43-3
Analyte or Component Names Dibutyl Sebacate
Shipping Condition Room Temperature
Molecular Formula C18 H34 O4
Certifications/Compliance ISO 17034
Synonym Decanedioic acid, dibutyl ester (9CI), Sebacic acid, dibutyl ester (6CI,8CI), Bis(n-butyl) sebacate, DBS, Di-n-Butyl sebacate, Dibutyl decanedioate, Dibutyl sebacate, Ergoplast SDB, Kodaflex DBS, NSC 3893, PX 404, Polycizer DBS, Reomol DBS, Sebacic acid di-n-butyl ester, Staflex DBS, Uniflex DBS
InChI Formula InChI=1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC
IUPAC Name dibutyl decanedioate
Molecular Weight (g/mol) 314.46
Quantity 500 mg
Formula Weight 314.2457 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.