missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Sodium, LoGiCal

Product Code. 30458660
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458660 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458660 Supplier LoGiCal Supplier No. LGCFOR0006.00
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac sodium
CAS 15307-79-6
Analyte or Component Names Diclofenac sodium
Shipping Condition Room Temperature
Molecular Formula C14 H10 Cl2 N O2 . Na
Certifications/Compliance ISO 9001
Synonym Diclofenac Sodium Assay Standard, Aceclofenac Imp. A (EP), Diclofenac Sodium, 2-[(2,6-dichlorophenyl)amino]-benzeneacetic acid monosodium salt, [o-(2,6-dichloroanilino)phenyl]-acetic acid monosodium salt, 2-(2,6-Dichloroanilino)phenylacetic acid sodium salt, Allvoran, Assaren, Benfofen, Delphimix, Dichronic, Diclo-Phlogont, Diclo-Puren, Diclobenin, Diclofen SR 100, Diclofenac sodium, Diclofenac sodium salt, Diclofenac-Na Emulgel, Diclofenacsodium Emulgel, Diclophenac sodium, Diclord, Dicloreum, Diklovit, Dolobasan, Duravolten, Effekton, Evofenac, Feloran, Fortfen, GP 45840, Hyanalgese D, Inflaban, Kriplex, N-(2,6-Dichlorophenyl)-o-aminophenylacetic acid sodium salt, Naclof, Neriodin, Novapirina, Orthofen, Orthophen, Primofenac, Profenac, Prophenatin, Rhumalgan, SR 318B, Sodium [o-(2,6-dichloroanilino)phenyl]acetate, Sodium diclofenac, Sorelmon, Tsudohmin, Valetan, Voldal, Voltaren, Voltaren Ophtha, Voltaren Ophtha CD, Voltarol, Voveran, Xenid, [o-(2,6-Dichloroanilino)phenyl]acetic acid sodium salt, Aceclofenac Impurity A
InChI Formula InChI=1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1
Solution Type Neat
SMILES [Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 318.13
Quantity 10 mg
Formula Weight 316.9986 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.