missing translation for 'onlineSavingsMsg'
Learn More

Dicumarol, Mikromol™

Product Code. 18072660
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072660 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072660 Supplier Mikromol Supplier No. MM3051.00
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dicumarol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dicoumarol
CAS 66-76-2
Analyte or Component Names Dicoumarol
Shipping Condition Room Temperature
Molecular Formula C19 H12 O6
Certifications/Compliance ISO 17025
Synonym Coumarin, 3,3'-methylenebis[4-hydroxy-, 3,3'-Methylenebis[4-hydroxy-1,2-benzopyrone], 3,3'-Methylenebis[4-hydroxycoumarin], Acadyl, Acavyl, Antitrombosin, Baracoumin, Bis(4-hydroxycoumarin-3-yl)methane, Bis-3,3'-(4-hydroxycoumarinyl)methane, Bishydroxycoumarin, Cuma, Cumid, Di-4-hydroxy-3,3'-methylenedicoumarin, Dicoumal, Dicoumarin, Dicoumarol, Dicuman, Dicumarine, Dicumarol, Dicumol, Dufalone, Kumoran, Melitoxin, NC 034, NSC 17860, NSC 221570, NSC 41834, Temparin, Trombosan
InChI Formula InChI=1S/C19H12O6/c20-16-10-5-1-3-7-14(10)24-18(22)12(16)9-13-17(21)11-6-2-4-8-15(11)25-19(13)23/h1-8,20-21H,9H2
SMILES OC1=C(CC2=C(O)c3ccccc3OC2=O)C(=O)Oc4ccccc14
IUPAC Name 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
Molecular Weight (g/mol) 336.3
Quantity 250 mg
Formula Weight 336.0634 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.