missing translation for 'onlineSavingsMsg'
Learn More

Myelin PLP Antibody (PLP1/4259), FITC, R&D Systems™

Product Code. 18602189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602189 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602189 Supplier Novus Biologicals Supplier No. NBP308534F
  • ONLINE EXCLUSIVE PRICE
    707.99 €
    619.50 € / 100µL
    Save 88.49 €
    12% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Myelin PLP Monoclonal specifically detects Myelin PLP in Bovine/Human/Mouse/Rat/Reptile samples. It is validated for Flow Cytometry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin PLP
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PLP1/4259
Conjugate FITC
Formulation PBS
Gene Alias HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
Host Species Mouse
Immunogen Synthetic peptide GRGTKF that recognizes the C terminal region (aa272-277) of Myelin PLP protein. (Uniprot: P60201)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5354
Target Species Bovine, Human, Mouse, Rat, Reptile
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.