missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate), Mikromol™

Product Code. 17953456
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953456 100 mg 100mg
17963216 25 mg 25mg
30461866 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953456 Supplier Mikromol Supplier No. MM0039.04
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
CAS 16150-45-1
Analyte or Component Names Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
Shipping Condition Room Temperature
Molecular Formula C27 H24 O11
Certifications/Compliance ISO 17025
Synonym Sodium Cromoglicate Imp. B (EP), Cromolyn USP Related Compound B, Cromolyn USP RC B, Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate), Ethyl Cromoglicate, Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)bis(oxy)]bis(4-oxo-4H-chromene-2-carboxylate), Cromolyn Related Compound B, Sodium Cromoglicate Impurity B
InChI Formula InChI=1S/C27H24O11/c1-3-33-26(31)22-11-16(29)24-18(7-5-9-20(24)37-22)35-13-15(28)14-36-19-8-6-10-21-25(19)17(30)12-23(38-21)27(32)34-4-2/h5-12,15,28H,3-4,13-14H2,1-2H3
SMILES CCOC(=O)C1=CC(=O)c2c(OCC(O)COc3cccc4OC(=CC(=O)c34)C(=O)OCC)cccc2O1
IUPAC Name ethyl 5-[3-(2-ethoxycarbonyl-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Molecular Weight (g/mol) 524.47
Quantity 100 mg
Formula Weight 524.1319 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.