missing translation for 'onlineSavingsMsg'
Learn More

Unity Lab Services Digital Education Module for Thermo CO2 Incubation

Product Code. 31188981
Change view
Click to view available options
Duration:
1 Day
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration
31188981 1 Day
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31188981 Supplier Unity Lab Services Supplier No. TINCCO2FOF
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Digital learning module on Thermo CO2 incubators, covering operation, maintenance, troubleshooting, and assessment for users.

  • Concise digital module focused on Thermo CO2 incubators
  • Clear guidance on daily operation and optimal usage
  • Essential maintenance practices to improve reliability
  • Simple troubleshooting steps for common issues
  • Knowledge check with completion certificate
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) CO2 Incubator
Duration 1 Day
Product Type Service
Target Audience Employees/Users
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.