missing translation for 'onlineSavingsMsg'
Learn More

Digoxin 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30461165
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461165 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461165 Supplier LoGiCal Supplier No. LGCAMP0212.0001
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Digoxin 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Digoxin
CAS 20830-75-5
Analyte or Component Names Digoxin
Shipping Condition Ice Pack (-20°C)
Molecular Formula C41 H64 O14
Certifications/Compliance ISO 17034
Synonym Digoxin (8 CI), Cardigox, Cardiogoxin, Cardioxin, Cardixin, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one
InChI Formula InChI=1 S/C41H64O14/c1-19-36(47)28(42)15-34(50-19)54-38-21(3)52-35(17-30(38)44)55-37-20(2)51-33(16-29(37)43)53-24-8-10-39(4)23(13-24)6-7-26-27(39)14-31(45)40(5)25(9-11-41(26,40)48)22-12-32(46)49-18-22/h12,19-21,23-31,33-38,42-45,47-48 H,6-11,13-18H2,1-5H3/t19-,20-,21-,23-,24+,25-,26-,27+,28+,29+,30+,31-,33+,34+,35+,36-,37-,38-,39+,40+,41+/m1/s1
SMILES O[C@@]12 [C@@](C)([C@H](CC1)C3=CC(=O)OC3)[C@H](O)C[C@]4([C@]2(CC[C@]5([C@]4(C)CC[C@H](O[C@H]6 C[C@H](O)[C@H](O[C@H]7 C[C@H](O)[C@H](O[C@H]8 C[C@H](O)[C@H](O)[C@@H](C)O8)[C@@H](C)O7)[C@@H](C)O6)C5)[H])[H])[H]
IUPAC Name 3-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S,17 R)-3-[(2 R,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2 H-furan-5-one
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 780.94
Quantity 1 mL
Formula Weight 780.4296 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.