missing translation for 'onlineSavingsMsg'
Learn More

Diltiazem Hydrochloride 1.0 mg/ml in Acetonitrile (as free base), LoGiCal

Product Code. 30462106
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30462106 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30462106

Brand: LoGiCal LGCAMP0068.0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diltiazem HCl
CAS 33286-22-5
Analyte or Component Names Diltiazem HCl
Shipping Condition Room Temperature
Molecular Formula C22H27ClN2O4S
Certifications/Compliance ISO 17034
Synonym 1,5-Benzothiazepin-4(5H)-one, 3-(acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-, hydrochloride (1:1), (2S,3S)-, 1,5-Benzothiazepin-4(5H)-one, 3-(acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-, monohydrochloride, (2S,3S)- (9CI), 1,5-Benzothiazepin-4(5H)-one, 3-(acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-, monohydrochloride, (2S-cis)-, (+)-Diltiazem hydrochloride, (2S,3S)-(+)-cis-Diltiazem hydrochloride, (S,S)-Diltiazem hydrochloride, Adizem, Altiazem, Anginyl, Angipress, Angizem, Bi-Teldiem, Blocalcin 60, Britiazim, Bruzem, CRD 401, Calcicard, Cardizem, Cardizem CD, Cardizem SR, Citizem, Cormax, Deltazen, Dicard, Dilacor, Dilacor XR, Diladel, Dilcardia, Dilgard, Dilpral, Dilrene, Diltiazem hydrochloride, Dilzacard, Dilzene, Herbesser, Kaizem, Kardil, Masdil, RG 83606, Tiazac, Tildiem, d-cis-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(p-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, d-cis-Diltiazem hydrochloride
Solution Type Single Solution
SMILES Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(=O)C
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 450.98
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.