missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Fumarate, Mikromol™

Product Code. 17943926
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943926 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943926 Supplier Mikromol Supplier No. MM3170.00
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethyl Fumarate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fumaric acid dimethyl ester
CAS 624-49-7
Analyte or Component Names Fumaric acid dimethyl ester
Shipping Condition Room Temperature
Molecular Formula C6 H8 O4
Certifications/Compliance ISO 17025
Synonym (2E)-2-Butenedioic Acid 1,4-Dimethyl Ester, (E)-2-Butenedioic Acid 1,4-Dimethyl Ester, trans-1,2-Ethylenedicarboxylic Acid, Fumaric Acid Dimethyl Ester, (2E)-2-Butenedioic Acid Dimethyl Ester, (E)-But-2-enedioic Acid Dimethyl Ester, AZL-O 211089, Allomaleic Acid Dimethyl Ester, BG 00012, Boletic Acid Dimethyl Ester, Dimethyl (E)-2-Butenedioate, Dimethyl (E)-Butenedioate, Dimethyl Fumarate, Dimethyl trans-Ethylenedicarboxylate, Methyl Fumarate, NSC 167432, NSC 25942, trans-1,2-Ethylenedicarboxylic Acid Dimethyl Ester, trans-Butenedioic Acid Dimethyl Ester, Fumaric acid dimethyl ester
InChI Formula InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+
SMILES COC(=O)\C=C\C(=O)OC
IUPAC Name dimethyl (E)-but-2-enedioate
Molecular Weight (g/mol) 144.13
Quantity 250 mg
Formula Weight 144.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.