missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl tetrachloroterephthalate, 97%, Thermo Scientific™

Product Code. 11352716
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352716 25 g 25g
11342716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352716 Supplier Thermo Scientific Alfa Aesar Supplier No. L08800.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1861-32-1
Molecular Formula C10H6Cl4O4
Molecular Weight (g/mol) 331.95
MDL Number MFCD00014902
InChI Key NPOJQCVWMSKXDN-UHFFFAOYSA-N
Synonym chlorthal-dimethyl, dacthal, dimethyl tetrachloroterephthalate, dacthalor, fatal, chlorthal-methyl, tetral, tetrachloroterephthalic acid dimethyl ester, dimethyl 2,3,5,6-tetrachloroterephthalate, dac 4
PubChem CID 2943
ChEBI CHEBI:34664
IUPAC Name dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl

Specifications

Melting Point 156°C to 159°C
Quantity 25 g
Beilstein 1888840
Merck Index 14,2839
Formula Weight 331.97
Percent Purity 97%
Chemical Name or Material Dimethyl tetrachloroterephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.