accessibility menu, dialog, popup

UserName

CH3OOClClClClOOCH3

CAS RN 1861-32-1

IUPAC Name: 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Synonyms: dacthal caswell no. 382 chlorthal dimethyl ester daktal dimethylester kyseliny tetrachlortereftalove chlorthal-methyl epa pesticide chemical code 078701 dac 4 tetral dacthalor
Molecular Weight (g/mol): 331.95
Molecular Formula: C10H6Cl4O4
InChi Key: NPOJQCVWMSKXDN-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (6)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (2)
Concentration 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results