missing translation for 'onlineSavingsMsg'
Learn More

Dioctadecyl 3,3'-Thiodipropionate, Mikromol

Product Code. 30457510
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457510 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457510 Supplier Mikromol Supplier No. MM3963.020100
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dioctadecyl 3,3'-Thiodipropionate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Plastic additive 17
CAS 693-36-7
Analyte or Component Names Plastic additive 17
Shipping Condition Room Temperature
Molecular Formula C42 H82 O4 S
Certifications/Compliance ISO 9001
Synonym 3,3'-Thiobispropanoic acid 1,1'-dioctadecyl ester, 3,3'-Thiobispropanoic acid dioctadecyl ester, 3,3'-Thiodipropionic acid dioctadecyl ester, Advastab 802, Advastab PS 802, Antiox S, Antioxidant 802, Arbestab DSTDP, Carstab DSTDP, Cyanox OTDP, Cyanox STDP, DSTDP, DSTP, DSTP Yoshitomi, Dioctadecyl 3,3'-thiodipropionate, Dioctadecyl thiodipropionate, Distearyl 3,3'-thiodipropionate, Distearyl thiodipropionate, Distearyl β,β'-thiodipropionate, Distearyl β-thiodipropionate, Distearyl-3,3-thiodipropionate, Evanstab 18, Hostanox SE 2, Hostanox SE 4, Hostanox VP-SE 2, Irganox 802FD, Irganox PS 802, Irganox PS 802FD, Irganox PS 802FL, Lasumit SG, Lowinox DSTDP, Lusmit SS, NSC 65493, Naugard DSTDP, Negonox DSTP, PS 802, PS 802FL, Plastanox STDP, Plastanox STDP Antioxidant, Seenox DS, Songnox DSTDP, Stearyl 3,3'-thiodipropionate, Sumilizer TPS, Thio 1, Varox DSTDP, Yoshinox DSTDP, Yoshinox DSTP
InChI Formula InChI=1S/C42H82O4S/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37-45-41(43)35-39-47-40-36-42(44)46-38-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3
Solution Type Neat
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC
IUPAC Name octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate
Molecular Weight (g/mol) 683.16
Quantity 100 mg
Formula Weight 682.5934 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.