missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312516
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312516 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312516 Supplier Dr. Ehrenstorfer Supplier No. DREA12881000AL100
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenhydramine hydrochloride
CAS 147-24-0
Analyte or Component Names Diphenhydramine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H21 N O . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Orphenadrine Hydrochloride Imp. D (EP), Orphenadrine Imp. D (EP), Diphenhydramine Hydrochloride, Orphenadrine Hydrochloride Imp. D (EP) as Hydrochloride, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8CI), b-Dimethylaminoethyl benzhydryl ether hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, Alledryl, Allergan, Allergina, Allergival, Amidryl, Asdrin, Bagodryl, Bax, Bena, Bena-Fedrin, Benadril hydrochloride, Benadryl, Benocten, Benodine, Benylin, Benzantin, Benzhydramine hydrochloride, Benzhydrol b-dimethylaminoethyl ether hydrochloride, Denydryl, Dibondrin, Difenhydramine hydrochloride, Dimedrol, Dimedrol hydrochloride, Diphantine, Fenylhist, Halbmond, Histacyl, Noctomin, Resmin, Restamin, S 8, Sedopretten, Sekundal D, Syntedril, Vena, Wehydryl, Orphenadrine Hydrochloride Impurity D as Hydrochloride, Orphenadrine Impurity D as Hydrochloride
InChI Formula InChI=1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H
SMILES Cl.CN(C)CCOC(c1ccccc1)c2ccccc2
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 291.82
Quantity 1 mL
Formula Weight 291.139 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.