missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethanone (Benzophenone), Mikromol™

Product Code. 30460282
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460282 4 x 25 mg -
17943176 25 mg 25mg
17931136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460282 Supplier Mikromol Supplier No. MM0630.0100254AMP
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diphenylmethanone (Benzophenone), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzophenone
CAS 119-61-9
Analyte or Component Names Benzophenone
Shipping Condition Room Temperature
Molecular Formula C13 H10 O
Certifications/Compliance ISO 17025
Synonym Benzophenone, Diphenylmethanone, Hydrocodone Hydrogen Tartrate 2.5-Hydrate Imp. H (EP), Phenytoin Imp. A (EP), Phenytoin Sodium Imp. A (EP), Dimenhydrinate Imp. J (EP), Diphenhydramine Hydrochloride Imp. E (EP), Diphenhydramine Imp. E (EP), Hydrocodone Imp. H (EP), SKF-3353, Ph Eur Diphenhydramine HCl Impurity E, Dimenhydrinate Impurity J, Diphenhydramine Hydrochloride Impurity E, Hydrocodone Hydrogen Tartrate 2.5-Hydrate Impurity H, Phenytoin Impurity A, Phenytoin Sodium Impurity A, Diphenhydramine Impurity E, Hydrocodone Impurity H, Phenytoin Impurity A
InChI Formula InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
Solution Type Neat
SMILES O=C(c1ccccc1)c2ccccc2
IUPAC Name diphenylmethanone
Molecular Weight (g/mol) 182.22
Quantity 4 x 25 mg
Formula Weight 182.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.