missing translation for 'onlineSavingsMsg'
Learn More

DLC1 Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30490563 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30490563 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30490563 Supplier Novus Biologicals Supplier No. NBP335894V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DLC1 Polyclonal antibody specifically detects DLC1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLC1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ARHGAP7 STARD12 deleted in liver cancer 1 variant 2, deleted in liver cancer 1, Deleted in liver cancer 1 protein, DLC-1, FLJ21120, HP, HP protein, KIAA1723, p122-RhoGAP, rho GTPase-activating protein 7, Rho-GTPase-activating protein 7, Rho-type GTPase-activating protein 7, StARD12, StAR-related lipid transfer (START) domain containing 12, StAR-related lipid transfer protein 12, START domain-containing protein 12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 204-463 of human DLC1 (NP_079043.3).,, Sequence:, DAPKVNAVDTLNVKDIAPEKQLLNSAVIAQQRRKPDPPKDENERSTCNVVQNEFLDTPCTNRGLPLLKTDFGSCLLQPPSCPNGMSAENGLEKSGFSQHQNKSPPKVKAEDGMQCLQLKETLATQEPTDNQVRLRKRKEIREDRDRARLDSMVLLIMKLDQLDQDIENALSTSSSPSGTPTNLRRHVPDLESGSESGADTISVNQTRVNLSSDTESTDLPSSTPVANSGTKPKTTAIQGISEKEKAGKLTFWFCFLANLF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10395
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.