missing translation for 'onlineSavingsMsg'
Learn More

DLG7/HURP Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30502152 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30502152 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30502152 Supplier Novus Biologicals Supplier No. NBP337984AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DLG7/HURP Polyclonal antibody specifically detects DLG7/HURP in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLG7/HURP
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DAP-5, Discs large homolog 7, discs, large (Drosophila) homolog-associated protein 5, disks large-associated protein 5, DLG7, Hepatoma up-regulated protein, HURPDLG1, KIAA0008discs, large homolog 7 (Drosophila), large homolog 7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 547-846 of human DLG7/HURP (NP_055565.3).,, Sequence:, KNVFRKKVVSGIASKPKQDDAGRIAARNRLAAIKNAMRERIRQEECAETAVSVIPKEVDKIVFDAGFFRVESPVKLFSGLSVSSEGPSQRLGTPKSVNKAVSQSRNEMGIPQQTTSPENAGPQNTKSEHVKKTLFLSIPESRSSIEDAQCPGLPDLIEENHVVNKTDLKVDCLSSERMSLPLLAGGVADDINTNKKEGISDVVEGMELNSSITSQDVLMSSPEKNTASQNSILEEGETKISQSELFDNKSLTTECHLLDSPGLNCSNPFTQLERRHQEHARHISFGGNLITFSPLQPGEF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9787
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.