missing translation for 'onlineSavingsMsg'
Learn More

DLL3 Antibody (SC16.15) [Alexa Fluor™ 594] - Chimeric, Novus Biologicals™

Product Code. 30533126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30533126 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533126 Supplier Novus Biologicals Supplier No. NBP345128AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DLL3 Monoclonal antibody specifically detects DLL3 in Cynomolgus Monkey,Human,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLL3
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone SC16.15
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3
Host Species Rabbit
Immunogen Three strains of mice (Balb/c, CD-1 and FVB) were immunized with recombinant human DLL3-Fc or with human DLL3-His fusion constructs (Uniprot#: Q9NYJ7)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10683
Target Species Cynomolgus Monkey, Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.