missing translation for 'onlineSavingsMsg'
Learn More

DNA Ligase I Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30502772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30502772 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30502772 Supplier Novus Biologicals Supplier No. NBP335879B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Ligase I Polyclonal antibody specifically detects DNA Ligase I in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase I
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 565-790 of human DNA Ligase I (NP_000225.1).,, Sequence:, CEYKYDGQRAQIHALEGGEVKIFSRNQEDNTGKYPDIISRIPKIKLPSVTSFILDTEAVAWDREKKQIQPFQVLTTRKRKEVDASEIQVQVCLYAFDLIYLNGESLVREPLSRRRQLLRENFVETEGEFVFATSLDTKDIEQIAEFLEQSVKDSCEGLMVKTLDVDATYEIAKRSHNWLKLKKDYLDGVGDTLDLVVIGAYLGRGKRAGRYGGFLLASYDEDSEEL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3978
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.