missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DNA Polymerase lambda Antibody [PE], Novus Biologicals™

Product Code. 30029659 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30029659 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30029659 Supplier Novus Biologicals™ Supplier No. NB10081665PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Polymerase lambda Polyclonal antibody specifically detects DNA Polymerase lambda in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Polymerase lambda
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias BETAN, DNA polymerase beta-2, DNA polymerase beta-N, DNA polymerase kappa, DNA polymerase lambda, EC 2.7.7.7, EC 4.2.99.-, FLJ46002, Pol beta2, Pol Lambda, POLKAPPA, polymerase (DNA directed), lambda
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human polymerase (DNA directed), lambda using the numbering given in entry NP_037406.1 (GeneID 27343).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, DNA Polymerases, DNA Repair, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27343
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.