Learn More
DNAJA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12236-1-AP-20UL
 View more versions of this product
Product Code. 16839833
  • 145.00 € / 20µL
Description
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.
Specifications
Specifications
DNAJA2
Polyclonal
Unconjugated
DNAJA2
CPR3, DJ3, DJA2, DNAJ, DNAJA2, DNJ3, HIRA interacting protein 4, HIRIP4, PRO3015, RDJ2
Rabbit
Antigen Affinity Chromatography
RUO
10294, 56445, 84026
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35824, O60884, Q9QYJ0
Dnaja2
DNAJA2 Fusion Protein Ag2877
20 μL
Primary
Rat, Mouse, Human