missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | DNAJA2 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16839833
|
Proteintech
12236-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16829833
|
Proteintech
12236-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.Specifications
| DNAJA2 | |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O35824, O60884, Q9QYJ0 | |
| 10294, 56445, 84026 | |
| DNAJA2 Fusion Protein Ag2877 | |
| Primary | |
| -20°C | |
| DNAJA2 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CPR3, DJ3, DJA2, DNAJ, DNAJA2, DNJ3, HIRA interacting protein 4, HIRIP4, PRO3015, RDJ2 | |
| Dnaja2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title