missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC12 Recombinant Rabbit Monoclonal Antibody (23GB1815), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553991
This item is not returnable.
View return policy
Description
This gene encodes a member of a subclass of the HSP40/DnaJ protein family. Members of this family of proteins are associated with complex assembly, protein folding, and export. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| DNAJC12 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DNAJC12 | |
| DnaJ (Hsp40) homolog, subfamily C, member 12; DnaJ heat shock protein family (Hsp40) member C12; dnaJ homolog subfamily C member 12; DnaJ subfamily C member 12-like protein; DNAJC12; DnaJ-like protein subfamily C member 12-like protein; J domain containing protein 1; J domain containing protein 1 (JDP1); J domain protein 1; j domain-containing protein 1; JDP1; mJDP1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56521 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1815 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UKB3 | |
| DNAJC12 | |
| A synthesized peptide derived from human DKC1 (140-198AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction