missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC12 Recombinant Rabbit Monoclonal Antibody (23GB1815), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | DNAJC12 |
|---|---|
| Clone | 23GB1815 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088311
![]() |
Invitrogen™
MA553990 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089577
|
Invitrogen™
MA553991 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJC12 Recombinant Monoclonal antibody specifically detects DNAJC12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| DNAJC12 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UKB3 | |
| 56521 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1815 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| DnaJ (Hsp40) homolog, subfamily C, member 12; DnaJ heat shock protein family (Hsp40) member C12; dnaJ homolog subfamily C member 12; DnaJ subfamily C member 12-like protein; DNAJC12; DnaJ-like protein subfamily C member 12-like protein; J domain containing protein 1; J domain containing protein 1 (JDP1); J domain protein 1; j domain-containing protein 1; JDP1; mJDP1 | |
| A synthesized peptide derived from human DKC1 (140-198AA). | |
| Primary | |
| -20°C | |
| DNAJC12 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title