missing translation for 'onlineSavingsMsg'
Learn More

DNAJC19 Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™

Product Code. 30488791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488791 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488791 Supplier Novus Biologicals Supplier No. NBP338125AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJC19 Polyclonal antibody specifically detects DNAJC19 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC19
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 19, DnaJ homolog subfamily C member 19, homolog of yeast TIM14, mitochondrial import inner membrane translocase subunit TIM14, Tim14, TIMM14TIM14Pam18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-116 of human DNAJC19 (NP_660304.1).,, Sequence:, MASTVVAVGLTIAAAGFAGRYVLQAMKHMEPQVKQVFQSLPKSAFSGGYYRGGFEPKMTKREAALILGVSPTANKGKIRDAHRRIMLLNHPDKGGSPYIAAKINEAKDLLEGQAKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 131118
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.