missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Domiphen Bromide, Mikromol™
Description
Domiphen Bromide
Specifications
Specifications
| Chemical Name or Material | Domiphen Bromide |
| CAS | 538-71-6 |
| Analyte or Component Names | Domiphen Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H40 N O . Br |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1-Dodecanaminium, N,N-dimethyl-N-(2-phenoxyethyl)-, bromide, Ammonium, dodecyldimethyl(2-phenoxyethyl)-, bromide, Dodecyldimethyl(2-phenoxyethyl)ammonium bromide, Bradonit, Bradoral, Bradosol, Bradosol bromide, Domiphen bromide, Fungitex, Fungitex R, Modicare, N-Dodecyl-N-(2-phenoxyethyl)dimethylammonium bromide, NSC 39415, Neo-Bradoral, Oradol, PDDB, Phenododecinium bromide, (beta-Phenoxyethyl)dimethyldodecylammonium bromide |
| InChI Formula | InChI=1S/C22H40NO.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-23(2,3)20-21-24-22-17-14-13-15-18-22;/h13-15,17-18H,4-12,16,19-21H2,1-3H3;1H/q+1;/p-1 |
| SMILES | [Br-].CCCCCCCCCCCC[N+](C)(C)CCOc1ccccc1 |
| IUPAC Name | dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?