missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dothiepin.HCl (cis/trans), Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Dothiepin.HCl (cis/trans)
Specifications
Specifications
| Chemical Name or Material | Dosulepin Hydrochloride |
| CAS | 897-15-4 |
| Analyte or Component Names | Dosulepin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N S . Cl H |
| Synonym | Dothiepin Hydrochloride, Dosulepin Hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN(C)CC\C=C\1/c2ccccc2CSc3ccccc13 |
| IUPAC Name | (3 E)-3-(6 H-benzo[c][1 ]benzothiepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction