missing translation for 'onlineSavingsMsg'
Learn More

Doxazosin EP Impurity E, Mikromol™

Product Code. 30049511
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049511 25 mg 25mg
30052364 100 mg 100mg
30464094 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049511 Supplier Mikromol Supplier No. MM0085.060025
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxazosin EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
CAS 27631-29-4
Analyte or Component Names 2,4-Dichloro-6,7-dimethoxyquinazoline
Shipping Condition Room Temperature
Molecular Formula C10 H8 Cl2 N2 O2
Certifications/Compliance ISO 17025
Synonym Doxazosin Mesilate Imp. E (EP), Doxazosin USP Related Compound E, Doxazosin USP RC E, 2,4-Dichloro-6,7-dimethoxyquinazoline, Doxazosin Related Compound E, Doxazosin Mesilate Impurity E, Doxazosin Impurity E
InChI Formula InChI=1S/C10H8Cl2N2O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(12)14-9(5)11/h3-4H,1-2H3
SMILES COc1cc2nc(Cl)nc(Cl)c2cc1OC
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
Molecular Weight (g/mol) 259.09
Quantity 25 mg
Formula Weight 257.9963 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.