missing translation for 'onlineSavingsMsg'
Learn More

Dronedarone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30458136
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458136 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458136 Supplier LoGiCal Supplier No. LGCAMP1294.0001
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Dronedarone Hydrochloride
CAS 141625-93-6
Analyte or Component Names Dronedarone Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C31 H44 N2 O5 S . Cl H
Certifications/Compliance ISO 17034
Synonym Methanesulfonamide, N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]-, hydrochloride (1:1), Methanesulfonamide, N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]-, monohydrochloride (9CI), Dronedarone hydrochloride, Multaq
InChI Formula InChI=1S/C31H44N2O5S.ClH/c1-5-8-12-29-30(27-23-25(32-39(4,35)36)15-18-28(27)38-29)31(34)24-13-16-26(17-14-24)37-22-11-21-33(19-9-6-2)20-10-7-3;/h13-18,23,32H,5-12,19-22H2,1-4H3;1H
Solution Type Single Solution
SMILES Cl.CCCCN(CCCC)CCCOc1ccc(cc1)C(=O)c2c(CCCC)oc3ccc(NS(=O)(=O)C)cc23
IUPAC Name N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 593.22
Quantity 1 mL
Formula Weight 592.2738 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.