missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 Antibody (4A4C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37309
This item is not returnable.
View return policy
Description
DRP1 Monoclonal specifically detects DRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| DRP1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| DNM1L | |
| Purified recombinant fragment of human DNM1L (AA: 69-213) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4A4C7 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
| Mouse | |
| 81.9 kDa | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
| 10059 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction