missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 Antibody (4A4C7), Novus Biologicals™
Mouse Monoclonal Antibody
217.00 € - 582.00 €
Spezifikation
| Antigen | DRP1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18654356
|
Novus Biologicals
NBP2-37309-0.025ml |
0.025 mL |
217.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18191818
|
Novus Biologicals
NBP2-37309 |
0.1 mL |
582.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
DRP1 Monoclonal specifically detects DRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Spezifikation
| DRP1 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
| Ascites with 0.03% Sodium Azide | |
| 10059 | |
| Purified recombinant fragment of human DNM1L (AA: 69-213) expressed in E. Coli. | |
| Primary | |
| 81.9 kDa |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat | |
| DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
| DNM1L | |
| IgG1 | |
| Unpurified |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts