missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DTD1 Polyclonal antibody specifically detects DTD1 in Human samples. It is validated for Immunoprecipitation
Specifications
Specifications
| Antigen | DTD1 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | bA379J5.3, bA555E18.1, C20orf88HARS2, chromosome 20 open reading frame 88, DNA-unwinding element-binding protein, D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae), D-tyrosyl-tRNA(Tyr) deacylase 1, DUEB, EC 3.1, Histidyl-tRNA synthase-related, histidyl-tRNA synthetase 2, MGC119131, MGC41905, pqn-68 |
| Host Species | Rabbit |
| Immunogen | Between 125 and 175 |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?