missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duloxetine.HCl, Lipomed
Descripción
Duloxetine.HCl
Especificaciones
Especificaciones
| Chemical Name or Material | Duloxetine hydrochloride |
| CAS | 136434-34-9 |
| Analyte or Component Names | Duloxetine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N O S . Cl H |
| Synonym | Duloxetine Hydrochloride, (S)-(+)-Duloxetine hydrochloride, (S)-(+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, (S)-Duloxetine hydrochloride, Cymbalta, Duloxetine hydrochloride, LY 248686 HCl, (gammaS)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (S)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (gammaS)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride (1:1) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CNCC[C@H](Oc1cccc2ccccc12)c3cccs3 |
| IUPAC Name | (3 S)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-2-ylpropan-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?