missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dyclonine Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Dyclonine Hydrochloride |
| CAS | 536-43-6 |
| Analyte or Component Names | Dyclonine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H27 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dyclonine Hydrochloride, 1-Propanone, 1-(4-butoxyphenyl)-3-(1-piperidinyl)-, hydrochloride (1:1), 1-Propanone, 1-(4-butoxyphenyl)-3-(1-piperidinyl)-, hydrochloride (9CI), Propiophenone, 4'-butoxy-3-piperidino-, hydrochloride (7CI,8CI), 4'-Butoxy-3-piperidinopropiophenone hydrochloride, Diclonina, Diclonine, Dyclocaine hydrochloride, Dyclone, Dyclonine, Dyclonine hydrochloride, Dyclothane, Tanaclone |
| InChI Formula | InChI=1S/C18H27NO2.ClH/c1-2-3-15-21-17-9-7-16(8-10-17)18(20)11-14-19-12-5-4-6-13-19;/h7-10H,2-6,11-15H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCCOc1ccc(cc1)C(=O)CCN2CCCCC2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction