missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dynactin 6 Polyclonal specifically detects Dynactin 6 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | Dynactin 6 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | dynactin 6, dynactin subunit 6, Dynactin subunit p27, P27, Protein WS-3, WS-3, WS3novel RGD-containing protein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Dynactin 6 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?