missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dysferlin Monoclonal antibody specifically detects Dysferlin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Dysferlin |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL10888 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | dysferlin, dysferlin, limb girdle muscular dystrophy 2B (autosomal recessive), dystrophy-associated fer-1-like 1, FER1L1Dystrophy-associated fer-1-like protein, Fer-1-like protein 1, FLJ00175, FLJ90168, LGMD2B |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from O75923, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?