Learn More
E-Cadherin Antibody (7H12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
Brand:  Novus Biologicals NBP2-19051
 View more versions of this product
Product Code. 18770204
  • 663.00 € / 0.10mL
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human, Mouse, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000
135 kDa