missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (7H12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
252.00 € - 663.00 €
Specifications
| Form | Ascites |
|---|---|
| Gene Symbols | CDH1 |
| Isotype | IgG1 |
| Molecular Weight of Antigen | 135 kDa |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18716493
|
Novus Biologicals
NBP2-19051SS |
252.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18770204
|
Novus Biologicals
NBP2-19051 |
663.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human, Mouse, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Ascites | |
| CDH1 | |
| IgG1 | |
| 135 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title