missing translation for 'onlineSavingsMsg'
Learn More

E-Cadherin Antibody (CL1172) [Alexa Fluor™ 594], Novus Biologicals™

Product Code. 30526091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30526091 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30526091 Supplier Novus Biologicals Supplier No. NBP343888AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen E-Cadherin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL1172
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P12830, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 999
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.