missing translation for 'onlineSavingsMsg'
Learn More

(E/Z)-Ferulic acid 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30312104
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312104 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312104 Supplier Dr. Ehrenstorfer Supplier No. DREA13644090AC1000
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ferulic Acid
CAS 1135-24-6
Analyte or Component Names Ferulic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H10 O4
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym (2E)-3-(4-Hydroxy-3-methoxyphenyl)-2-acrylic acid, (2E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid, (2E)-3-(4-Hydroxy-3-methoxyphenyl)prop-2-enoic acid, (E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid, (E)-4-Hydroxy-3-methoxycinnamic acid, (E)-Ferulic acid, Fumalic acid, trans-4-Hydroxy-3-methoxycinnamic acid, trans-Ferulic acid
InChI Formula InChI=1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)
SMILES COc1cc(C=CC(=O)O)ccc1O
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
Solvent or Matrix Acetone
Molecular Weight (g/mol) 194.18
Quantity 1 mL
Formula Weight 194.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.