missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NB100-1869SS
This item is not returnable.
View return policy
Description
EAAT1/GLAST-1/SLC1A3 Polyclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
| EAAT1/GLAST-1/SLC1A3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
| Rabbit | |
| 60 kDa | |
| 0.025 mL | |
| Cellular Markers, Cognition and Behavior | |
| 6507 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 μg/mL, Flow Cytometry 1:200-1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular) 1:500 | |
| P24942 | |
| SLC1A3 | |
| A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [Uniprot: P24942] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat and mouse. Based upon 100% immunogen sequence similarity, this antibody is predicted to cross-react with Bovine also. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction